Learn More
Strem, An Ascensus Company CAS 1022915-09-8. 50mg. (11bR11bR)-22-[Oxybis(methylene)]bis[4-hydroxy-44-dioxide-dinaphtho[21-d12-f][132]dioxaphosphepin] 98% (99% ee).
SDP
Catalog No. 502110730
Encompass
CAS 1022915-09-8. 50mg. (11bR11bR)-22-[Oxybis(methylene)]bis[4-hydroxy-44-dioxide-dinaphtho[21-d12-f][132]dioxaphosphepin] 98% (99% ee). . Molecular Weight 738.6. Molecular Formula C42H28O9P2. Color/form white to light yellow pwdr. Strem 15-0354. http//www.strem.com/catalog/v/15-0354/
Catalog No. 50-211-0730 Supplier Strem, An Ascensus Company Supplier No. 15035450MG
CAS 1022915-09-8. 50mg. (11bR11bR)-22-[Oxybis(methylene)]bis[4-hydroxy-44-dioxide-dinaphtho[21-d12-f][132]dioxaphosphepin] 98% (99% ee). . Molecular Weight 738.6. Molecular Formula C42H28O9P2. Color/form white to light yellow pwdr. Strem 15-0354. http//www.strem.com/catalog/v/15-0354/