Learn More
Strem, An Ascensus Company CAS . 50mg. (11bS)-26-Bis[4-(2-naphthalenyl)phenyl]-89101112131415-octahydro-4-hydroxy-4-oxide-dinaphtho[21-d12-f][132]dioxaphosphepin 98% (99% ee).
SDP
Catalog No. 502110731
Encompass
50mg. (11bS)-26-Bis[4-(2-naphthalenyl)phenyl]-89101112131415-octahydro-4-hydroxy-4-oxide-dinaphtho[21-d12-f][132]dioxaphosphepin 98% (99% ee). . Molecular Weight 760.9. Molecular Formula C52H41O4P. Color/form white to light yellow pwdr. Strem 15-0390. http//www.strem.com/catalog/v/15-0390/
Catalog No. 50-211-0731 Supplier Strem, An Ascensus Company Supplier No. 15039050MG
50mg. (11bS)-26-Bis[4-(2-naphthalenyl)phenyl]-89101112131415-octahydro-4-hydroxy-4-oxide-dinaphtho[21-d12-f][132]dioxaphosphepin 98% (99% ee). . Molecular Weight 760.9. Molecular Formula C52H41O4P. Color/form white to light yellow pwdr. Strem 15-0390. http//www.strem.com/catalog/v/15-0390/