Learn More
Chiral Technologies, Inc. CHIRALCEL OD
Catalog No. NC9297514
Encompass
CHIRALCEL OD 10micrometer Particle Size 4.6 x 250mm Product Type: Analytical Adsorbent: 3 5-dimethylphenyl carbamate
Catalog No. NC9297514 Supplier Chiral Technologies, Inc. Supplier No. 14025
CHIRALCEL OD 10micrometer Particle Size 4.6 x 250mm Product Type: Analytical Adsorbent: 3 5-dimethylphenyl carbamate