Learn More
Chiral Technologies, Inc. CHIRALCEL OD-H
Catalog No. NC9874347
Encompass
CHIRALCEL OD-H 5micrometer Particle Size 4.6 x 250mm Product Type: Analytical Adsorbent: Cellulose tris (3 5-dimethylphenyl carbamate)
Catalog No. NC9874347 Supplier Chiral Technologies, Inc. Supplier No. 14325
CHIRALCEL OD-H 5micrometer Particle Size 4.6 x 250mm Product Type: Analytical Adsorbent: Cellulose tris (3 5-dimethylphenyl carbamate)