Learn More
Chiral Technologies, Inc. CHIRALCEL OJ-3 4.6X150MM 3 MIC
Catalog No. NC0243766
Encompass
Coated Polysaccharid CHIRALCEL OJ-3, 3 micron, 4.6 mm ID x 150 mm LN HPLC HPLC analytical column
Catalog No. NC0243766 Supplier Chiral Technologies, Inc. Supplier No. 17524
Coated Polysaccharid CHIRALCEL OJ-3, 3 micron, 4.6 mm ID x 150 mm LN HPLC HPLC analytical column