Learn More
Chiral Technologies, Inc. CHIRALCEL OJ-3R High-Performance HPLC Column, 3 µm Particle Size, Fast Analysis
Catalog No. NC3711099
Encompass
The CHIRALCEL OJ-3R is a high-performance HPLC analytical column designed for effective chiral separation of enantiomers in reversed-phase applications. It offers fast analysis, high resolution, and compatibility with various HPLC and UHPLC systems. Made with a cellulose tris(4-methylbenzoate) chiral selector and a 3 µm silica gel support, it excels in analyzing biological and water-soluble samples, providing direct method transfer between different column sizes for enhanced versatility.
Catalog No. NC3711099 Supplier Chiral Technologies, Inc. Supplier No. 17894
The CHIRALCEL OJ-3R is a high-performance HPLC analytical column designed for effective chiral separation of enantiomers in reversed-phase applications. It offers fast analysis, high resolution, and compatibility with various HPLC and UHPLC systems. Made with a cellulose tris(4-methylbenzoate) chiral selector and a 3 µm silica gel support, it excels in analyzing biological and water-soluble samples, providing direct method transfer between different column sizes for enhanced versatility.