Learn More
Chiral Technologies, Inc. CHIRALCEL OX-3 HPLC Column, 3 µm Particle Size, 4.6 mm Internal Diameter, 250 mm Length
Catalog No. NC3343700
Encompass
The CHIRALCEL OX-3 is an advanced analytical column designed for high-performance liquid chromatography (HPLC) and supercritical fluid chromatography (SFC). With a 4.6 mm internal diameter and 250 mm length, it features a 3 µm particle size that enables rapid and efficient separation of optical isomers, making it an invaluable tool in various enantiomeric applications. Its superior resolving power and compatibility with a range of solvent systems set it apart from traditional columns, delivering high efficiency in chiral separations.
Catalog No. NC3343700 Supplier Chiral Technologies, Inc. Supplier No. 63525
The CHIRALCEL OX-3 is an advanced analytical column designed for high-performance liquid chromatography (HPLC) and supercritical fluid chromatography (SFC). With a 4.6 mm internal diameter and 250 mm length, it features a 3 µm particle size that enables rapid and efficient separation of optical isomers, making it an invaluable tool in various enantiomeric applications. Its superior resolving power and compatibility with a range of solvent systems set it apart from traditional columns, delivering high efficiency in chiral separations.