Learn More
Chiral Technologies, Inc. CHIRALCEL OZ-3R | 4.6mm x 100mm | 3 µm
Catalog No. NC1870575
Encompass
CHIRALCEL OZ-3R | 4.6mm x 100mm | 3 µm | Compatible with HPLC (Normal Phase and Reversed-phase) and SFC
Catalog No. NC1870575 Supplier Chiral Technologies, Inc. Supplier No. 42823
CHIRALCEL OZ-3R | 4.6mm x 100mm | 3 µm | Compatible with HPLC (Normal Phase and Reversed-phase) and SFC