Learn More
Chiral Technologies, Inc. CHIRALCEL OZ3 LC 150X4.6 3UM P
Catalog No. NC0207943
Encompass
Coated Polysaccharide, CHIRALCEL OZ-3, 3 micron,4.6 mm ID x 150mm LN, analytical column
Catalog No. NC0207943 Supplier Chiral Technologies, Inc. Supplier No. 42524
Coated Polysaccharide, CHIRALCEL OZ-3, 3 micron,4.6 mm ID x 150mm LN, analytical column