Learn More
Chiral Technologies, Inc. CHIRALPAK IF 4.6x100mm, 5 micron. Compatible with HPLC (Normal Phase and Reversed-Phase) and SFC
Catalog No. NC3433425
Encompass
CHIRALPAK IF 4.6x100mm, 5 micron. Compatible with HPLC (Normal Phase and Reversed-Phase) and SFC
Catalog No. NC3433425 Supplier Chiral Technologies, Inc. Supplier No. 86423
CHIRALPAK IF 4.6x100mm, 5 micron. Compatible with HPLC (Normal Phase and Reversed-Phase) and SFC