Learn More
Chiral Technologies, Inc. CHIRALPAK IC-3 HPLC Column, High Durability, Immobilized Polysaccharide Phase, 2.1 mm
Catalog No. NC2747371
Encompass
The CHIRALPAK IC-3 column is designed for efficient separation of enantiomers in HPLC applications. Its immobilized cellulose tris stationary phase and silica gel base provide exceptional solvent versatility and stability, enabling method development for compounds with limited solubility. The column can operate across a broad pH range and withstand high pressures, making it suitable for various chromatography techniques including HPLC and UHPLC. Its durability ensures reliable long-term operation for laboratory professionals.
Catalog No. NC2747371 Supplier Chiral Technologies, Inc. Supplier No. 83595
The CHIRALPAK IC-3 column is designed for efficient separation of enantiomers in HPLC applications. Its immobilized cellulose tris stationary phase and silica gel base provide exceptional solvent versatility and stability, enabling method development for compounds with limited solubility. The column can operate across a broad pH range and withstand high pressures, making it suitable for various chromatography techniques including HPLC and UHPLC. Its durability ensures reliable long-term operation for laboratory professionals.