Learn More
Chiral Technologies, Inc. CHIRALPAK AD-3 3 MICRON
Catalog No. NC9014435
Encompass
Daicel CHIRALPAK AD-3 HPLC Analytical Column, 3 μm, ID 4.6 mm x L 250 mm - 19525 CHIRALPAK® & CHIRALCEL® Coated phases.
Catalog No. NC9014435 Supplier Chiral Technologies, Inc. Supplier No. 19525
Daicel CHIRALPAK AD-3 HPLC Analytical Column, 3 μm, ID 4.6 mm x L 250 mm - 19525 CHIRALPAK® & CHIRALCEL® Coated phases.