Learn More
Chiral Technologies, Inc. CHIRALPAK AD-3 4.6X150 COLUMN
Catalog No. NC0216743
Encompass
Coated Polysaccharid CHIRALPAK AD-3, 3 micron, 4.6 mm ID x 150 mm LN HPLC HPLC analytical column
Catalog No. NC0216743 Supplier Chiral Technologies, Inc. Supplier No. 19524
Coated Polysaccharid CHIRALPAK AD-3, 3 micron, 4.6 mm ID x 150 mm LN HPLC HPLC analytical column