Learn More
Chiral Technologies, Inc. CHIRALPAK AD 4.6MM X 250MM
Catalog No. NC9520023
Encompass
CHIRALPAK AD Particle Size (micro): 10 Internal Diameter (mm): 4.6 Column Length (mm): 250 Product Type: Analytical Adsorbent: Amylose tris (3 5-dimethylphenyl carbamate)
Catalog No. NC9520023 Supplier Chiral Technologies, Inc. Supplier No. 19025
CHIRALPAK AD Particle Size (micro): 10 Internal Diameter (mm): 4.6 Column Length (mm): 250 Product Type: Analytical Adsorbent: Amylose tris (3 5-dimethylphenyl carbamate)