Learn More
Chiral Technologies, Inc. HPLC Column CHIRALPAK AD-H, 250 x 4.6 mm, 5 um
Catalog No. NC1730874
Encompass
Chiralpak Chiral Column Amylose, Particle Size: 5 um, Inner Diameter: 4.6mm, Length: 250mm.
Catalog No. NC1730874 Supplier Chiral Technologies, Inc. Supplier No. 19325
Chiralpak Chiral Column Amylose, Particle Size: 5 um, Inner Diameter: 4.6mm, Length: 250mm.