Learn More
Chiral Technologies, Inc. CHIRALPAK AD-RH 150X4.6
Catalog No. NC9733634
Encompass
CHIRALPAK AD-RH Particle Size (micro): 5 Internal Diameter (mm): 4.6 Column Length (mm): 150 Product Type: Analytical Adsorbent: Amylose tris (3 5-dimethylphenyl carbamate) Applications: Reversed Phase type of CHIRALCEL AD-H
Catalog No. NC9733634 Supplier Chiral Technologies, Inc. Supplier No. 19724
CHIRALPAK AD-RH Particle Size (micro): 5 Internal Diameter (mm): 4.6 Column Length (mm): 150 Product Type: Analytical Adsorbent: Amylose tris (3 5-dimethylphenyl carbamate) Applications: Reversed Phase type of CHIRALCEL AD-H