Learn More
Chiral Technologies, Inc. Chiralpak AS-3R 3 µm 4.6x100mm
Supplier:  Chiral Technologies, Inc. 20823
Encompass
Coated Analytical Column. Reversed phase only. Compatible with HPLC and SFC.
Catalog No. NC1656748