Learn More
Chiral Technologies, Inc. CHIRALPAK AS-H 4.6X150MM 5UM
Catalog No. NC9006764
Encompass
CHIRALPAK AS-H, Analytical Columns for HPLC and SFC, H Series, 4.6 dia. x 150mm L, 5micro
Catalog No. NC9006764 Supplier Chiral Technologies, Inc. Supplier No. 20324
CHIRALPAK AS-H, Analytical Columns for HPLC and SFC, H Series, 4.6 dia. x 150mm L, 5micro