Learn More
Chiral Technologies, Inc. CHIRALPAK AY-3 | 4.6mm x 100mm | 3 µm
Catalog No. NC1870574
Encompass
CHIRALPAK AY-3 | 4.6mm x 100mm | 3 µm | Compatible with HPLC (Normal Phase and Reversed-phase) and SFC
Catalog No. NC1870574 Supplier Chiral Technologies, Inc. Supplier No. 47523
CHIRALPAK AY-3 | 4.6mm x 100mm | 3 µm | Compatible with HPLC (Normal Phase and Reversed-phase) and SFC