Learn More
Chiral Technologies, Inc. HPLC Column CHIRALPAK AZ-3, 150 x 4.6 mm, 3 um
Supplier:  Chiral Technologies, Inc. 61524
Encompass
Chiralpak Chiral Column Amylose, Particle Size: 3 um, Inner Diameter: 4.6mm, Length: 150mm.
Catalog No. NC1510512