Learn More
Chiral Technologies, Inc. Chiralpak AZ-3R 3 µm 4.6x100mm
Catalog No. NC1656749
Encompass
Coated Analytical Column. Reversed phase only. Compatible with HPLC and SFC.
Catalog No. NC1656749 Supplier Chiral Technologies, Inc. Supplier No. 61823
Coated Analytical Column. Reversed phase only. Compatible with HPLC and SFC.