Learn More
Chiral Technologies, Inc. CHIRALPAK IB-3, HPLC COLUMN, 4
Catalog No. NC9002167
Encompass
Immobilized Polysaccaride, CHIRALPAK IB-3, 3 micron, 4.6 mm ID x 150mm LN Analytical Column
Catalog No. NC9002167 Supplier Chiral Technologies, Inc. Supplier No. 81524
Immobilized Polysaccaride, CHIRALPAK IB-3, 3 micron, 4.6 mm ID x 150mm LN Analytical Column