Learn More
Chiral Technologies, Inc. CHIRALPAK IB N-3 3µm Column, Analytical Column, Immobilized Polysaccharide Stationary Phase
Catalog No. NC1956675
Encompass
The CHIRALPAK IB N-3 4.6x250 mm, 3 µm column by CHIRAL TECHNOLOGIES INC is designed for analytical HPLC and SFC applications, enabling robust and flexible chiral separations. Its immobilized polysaccharide phase ensures high stability across various solvents, enhancing selectivity and productivity. With versatile operating modes and temperature ranges, this column delivers efficient performance for diverse analytical needs.
Catalog No. NC1956675 Supplier Chiral Technologies, Inc. Supplier No. 88525
Add to Cart
Edge
Add to Cart
The CHIRALPAK IB N-3 4.6x250 mm, 3 µm column by CHIRAL TECHNOLOGIES INC is designed for analytical HPLC and SFC applications, enabling robust and flexible chiral separations. Its immobilized polysaccharide phase ensures high stability across various solvents, enhancing selectivity and productivity. With versatile operating modes and temperature ranges, this column delivers efficient performance for diverse analytical needs.