Learn More
Chiral Technologies, Inc. CHIRALPAK IC 4.6 X 150MM 3UM
Catalog No. NC9840795
Encompass
Immobilized Polysaccaride, CHIRALPAK IC-3, 3 micron, 4.6 mm ID x 150mm LN Analytical Column
Catalog No. NC9840795 Supplier Chiral Technologies, Inc. Supplier No. 83524
Immobilized Polysaccaride, CHIRALPAK IC-3, 3 micron, 4.6 mm ID x 150mm LN Analytical Column