Learn More
Chiral Technologies, Inc. CHIRALPAK IC ANALYTICAL COLUMN
Catalog No. NC9560276
Encompass
Immobilized Polysaccharide, CHIRALPAK IC, 5 micron, 4.6 mm ID x 250 mm LN Analytical column
Catalog No. NC9560276 Supplier Chiral Technologies, Inc. Supplier No. 83325
Immobilized Polysaccharide, CHIRALPAK IC, 5 micron, 4.6 mm ID x 250 mm LN Analytical column