Learn More
Chiral Technologies, Inc. CHIRALPAK ID 4.6 X 150 MM (5UM
Catalog No. NC0159412
Encompass
Immobilized Polysaccharide, CHIRALPAK ID, 5 micron, 4.6 mm ID x 150 mm LN Analytical column
Catalog No. NC0159412 Supplier Chiral Technologies, Inc. Supplier No. 84324
Immobilized Polysaccharide, CHIRALPAK ID, 5 micron, 4.6 mm ID x 150 mm LN Analytical column