Learn More
Chiral Technologies, Inc. CHIRALPAK IF-3, 4.6X150MM, 3MI
Catalog No. NC0402430
Encompass
Immobilized Polysaccaride, CHIRALPAK IF-3, 3 micron, 4.6 mm ID x 250mm LN Analytical Column
Catalog No. NC0402430 Supplier Chiral Technologies, Inc. Supplier No. 86524
Immobilized Polysaccaride, CHIRALPAK IF-3, 3 micron, 4.6 mm ID x 250mm LN Analytical Column