Learn More
Chiral Technologies, Inc. HPLC Column CHIRALPAK IG, 250 x 4.6 mm, 5 um
Supplier:  Chiral Technologies, Inc. 87325
Encompass
Chiralpak Chiral Column Cellulose, Particle Size: 5 um, Inner Diameter: 4.6mm, Length: 250mm.
Catalog No. NC1140194