Learn More
Chiral Technologies, Inc. CHIRALPAK® IJ-3, 4.6mm x 150mm, 3 um
Catalog No. NC2754712
Encompass
CHIRALPAK® IJ-3 4.6x150mm, 3 micron. Compatible with HPLC (Normal Phase and Reversed-Phase) and SFC
Catalog No. NC2754712 Supplier Chiral Technologies, Inc. Supplier No. 90524
CHIRALPAK® IJ-3 4.6x150mm, 3 micron. Compatible with HPLC (Normal Phase and Reversed-Phase) and SFC