Learn More
Chiral Technologies, Inc. CHIRALPAK IK-3 Chiral HPLC Column, 150 mm Length, 4.6 mm Inner Diameter, 3 µm Particle Size
Catalog No. NC2541455
Encompass
The CHIRALPAK IK-3 is a highly versatile analytical chiral HPLC column designed for efficient enantiomer resolution. With a unique immobilized polysaccharide stationary phase, it offers excellent robustness and compatibility with various solvents, making it suitable for normal phase, reversed-phase, and supercritical fluid chromatography applications. Its exceptional durability and selectivity set it apart, ensuring reliable performance across a wide range of organic solvents.
Catalog No. NC2541455 Supplier Chiral Technologies, Inc. Supplier No. 91524
The CHIRALPAK IK-3 is a highly versatile analytical chiral HPLC column designed for efficient enantiomer resolution. With a unique immobilized polysaccharide stationary phase, it offers excellent robustness and compatibility with various solvents, making it suitable for normal phase, reversed-phase, and supercritical fluid chromatography applications. Its exceptional durability and selectivity set it apart, ensuring reliable performance across a wide range of organic solvents.