Learn More
Chiral Technologies, Inc. Chiralpak IM/SFC Column, Immobilized Cellulose, Excellent Chemical Stability, 21 mm
Catalog No. NC3366774
Encompass
The CHIRALPAK IM/SFC 21X250MM 5MIC is a high-performance chiral chromatography column designed for Supercritical Fluid Chromatography (SFC) and HPLC applications. It offers enhanced robustness and universal solvent compatibility through its immobilized polysaccharide stationary phase, allowing for novel selectivity profiles and improved productivity in challenging enantiomeric separations. This combination of features differentiates it from standard chiral columns, making it a versatile tool for chromatographers.
Catalog No. NC3366774 Supplier Chiral Technologies, Inc. Supplier No. 92445
Add to Cart
Add to Cart
Request a Quote
The CHIRALPAK IM/SFC 21X250MM 5MIC is a high-performance chiral chromatography column designed for Supercritical Fluid Chromatography (SFC) and HPLC applications. It offers enhanced robustness and universal solvent compatibility through its immobilized polysaccharide stationary phase, allowing for novel selectivity profiles and improved productivity in challenging enantiomeric separations. This combination of features differentiates it from standard chiral columns, making it a versatile tool for chromatographers.