Learn More
Chiral Technologies, Inc. CHIRALPAK ZWIX(+) Analytical Column, Zwitterionic Stationary Phase, Compatible with HPLC and SFC, 3.0 mm
Catalog No. NC2115854
Encompass
The CHIRALPAK ZWIX is a chiral analytical column designed for high-performance separations of zwitterionic compounds. With its zwitterionic stationary phase and advanced amphoteric ion exchange mechanism, it allows for efficient enantioseparation, notably for underivatized amino acids and small peptides. Its versatility with various mobile phases and compatibility with HPLC and SFC techniques make it ideal for diverse analytical applications. Enhanced separation efficiency is achieved with a 3 µm particle size, ensuring superior results.
This item is not returnable. View return policy
Catalog No. NC2115854 Supplier Chiral Technologies, Inc. Supplier No. 51584
The CHIRALPAK ZWIX is a chiral analytical column designed for high-performance separations of zwitterionic compounds. With its zwitterionic stationary phase and advanced amphoteric ion exchange mechanism, it allows for efficient enantioseparation, notably for underivatized amino acids and small peptides. Its versatility with various mobile phases and compatibility with HPLC and SFC techniques make it ideal for diverse analytical applications. Enhanced separation efficiency is achieved with a 3 µm particle size, ensuring superior results.