Learn More
Strem, An Ascensus Company 25mg. (11aS)-3,7-Di-9-anthracenyl-10,11,12,13-tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 95%, (99% ee).
SDP
Catalog No. 502065474
Encompass
25mg. (11aS)-3,7-Di-9-anthracenyl-10,11,12,13-tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 95%, (99% ee). . Molecular Weight: 722.8. Molecular Formula: C49H39O4P. Color/form: off white to light yellow pwdr. Strem# 15-8570. http://www.strem.com/catalog/v/15-8570/
Catalog No. 50-206-5474 Supplier Strem, An Ascensus Company Supplier No. 15857025MG
25mg. (11aS)-3,7-Di-9-anthracenyl-10,11,12,13-tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 95%, (99% ee). . Molecular Weight: 722.8. Molecular Formula: C49H39O4P. Color/form: off white to light yellow pwdr. Strem# 15-8570. http://www.strem.com/catalog/v/15-8570/