Learn More
Strem, An Ascensus Company 100mg. (11aS)-3,7-Bis[3,5-bis[3,5-bis(trifluoromethyl)phenyl]phenyl]-10,11,12,13-tetrahydro-5-hydroxy-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee).
SDP
Catalog No. 502065415
Encompass
100mg. (11aS)-3,7-Bis[3,5-bis[3,5-bis(trifluoromethyl)phenyl]phenyl]-10,11,12,13-tetrahydro-5-hydroxy-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee). . Molecular Weight: 1314.8. Molecular Formula: C61H31F24O4P. Color/form: off white to light yellow pwdr. Strem# 15-0564. http://www.strem.com/catalog/v/15-0564/
Catalog No. 50-206-5415 Supplier Strem, An Ascensus Company Supplier No. 150564100MG
100mg. (11aS)-3,7-Bis[3,5-bis[3,5-bis(trifluoromethyl)phenyl]phenyl]-10,11,12,13-tetrahydro-5-hydroxy-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee). . Molecular Weight: 1314.8. Molecular Formula: C61H31F24O4P. Color/form: off white to light yellow pwdr. Strem# 15-0564. http://www.strem.com/catalog/v/15-0564/