Learn More
Strem, An Ascensus Company 25mg. (11aS)-3,7-Bis[3,5-bis(tert-butyl)phenyl]-10,11,12,13-tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee).
SDP
Catalog No. 502065497
Encompass
25mg. (11aS)-3,7-Bis[3,5-bis(tert-butyl)phenyl]-10,11,12,13-tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee). . Molecular Weight: 747. Molecular Formula: C49H63O4P. Color/form: off white to light yellow pwdr. Strem# 15-8593. http://www.strem.com/catalog/v/15-8593/
Catalog No. 50-206-5497 Supplier Strem, An Ascensus Company Supplier No. 15859325MG
25mg. (11aS)-3,7-Bis[3,5-bis(tert-butyl)phenyl]-10,11,12,13-tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee). . Molecular Weight: 747. Molecular Formula: C49H63O4P. Color/form: off white to light yellow pwdr. Strem# 15-8593. http://www.strem.com/catalog/v/15-8593/