Learn More
Strem, An Ascensus Company 25mg. (11aR)-10,11,12,13-Tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-3,7-di-2-naphthalenyl-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee).
SDP
Catalog No. 502065468
Encompass
25mg. (11aR)-10,11,12,13-Tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-3,7-di-2-naphthalenyl-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee). . Molecular Weight: 622.7. Molecular Formula: C41H35O4P. Color/form: off white to light yellow pwdr. Strem# 15-8566. http://www.strem.com/catalog/v/15-8566/
Catalog No. 50-206-5468 Supplier Strem, An Ascensus Company Supplier No. 15856625MG
25mg. (11aR)-10,11,12,13-Tetrahydro-10,10,13,13-tetramethyl-5-hydroxy-3,7-di-2-naphthalenyl-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin, 98%, (99% ee). . Molecular Weight: 622.7. Molecular Formula: C41H35O4P. Color/form: off white to light yellow pwdr. Strem# 15-8566. http://www.strem.com/catalog/v/15-8566/