Learn More
Regis Technologies Inc HPLC Column, Chiral, Regis, ChiroSil RCA (+), 150mm x 4.6mm, 5um, 3cm/5cm diameters avail.
SDP
Catalog No. 50442786
Encompass
ChiroSil is a ChiroSil Crown-Ether Chiral CSP. ChiroSil columns are ideal for the separation of chiral compounds containing almost all primary amino groups and natural and unnatural amino acids and their derivatives.This CSP offers excellent performance, high efficiency, and reproducibility. The ChiroSil stationary phase delivers long life and high reliability due to covalent bonding. Change the elution order if necessary by selecting ChiroSil RCA (+) or ChiroSil SCA (-) column. ChiroSil works with conventional and extended solvent ranges. Choose original ChiroSil columns for greater separation and resolution factors.Bulk quantity purchases can be provided.
Catalog No. 50-442-786 Supplier Regis Technologies Inc Supplier No. 1799001300
ChiroSil is a ChiroSil Crown-Ether Chiral CSP. ChiroSil columns are ideal for the separation of chiral compounds containing almost all primary amino groups and natural and unnatural amino acids and their derivatives.This CSP offers excellent performance, high efficiency, and reproducibility. The ChiroSil stationary phase delivers long life and high reliability due to covalent bonding. Change the elution order if necessary by selecting ChiroSil RCA (+) or ChiroSil SCA (-) column. ChiroSil works with conventional and extended solvent ranges. Choose original ChiroSil columns for greater separation and resolution factors.Bulk quantity purchases can be provided.