Learn More
Med Vet International Miltex Freer Chisel, Single-Cut, Blade 2mm Wide, Each, 62-40
Supplier:  Med Vet International MIL6240
Encompass_Preferred
Specialized rhinology tool that surgeons can use to cut and reshape the delicate bone surface of the nasal septum. Solid straight shank for higher surgical precision. Extremely sharp tip makes it impossible to incise and widen ridges. 2mm.
Catalog No. 50-282-8975