Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chitin

Catalog No. AAJ6120622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6120622 100 g
AAJ6120636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6120622 Supplier Thermo Scientific Chemicals Supplier No. J6120622
Add to Cart

missing translation for 'validMainframeMsg'

A long-chain polymer of a N-acetylglucosamine

Chitin is used in the analysis of Chitinase. It is plays a role in the formation of arthropod exoskeletons. Chitin can be processed further into different derivatives. The two main derivatives are chitosan and amino glucose. other applications in cosmetics include as carboxymethyl chitin (moisturizers, changes flow properties), antistatic effect due to cationic properties (hair products), as antibacterial and antifungal agents and for treatment of mucous membranes, in tumor therapies, in micro surgery, neurosurgery, for treatment of chronic wounds, ulcers and bleeding (chitin powder).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chitin is used in the analysis of Chitinase. It is plays a role in the formation of arthropod exoskeletons. Chitin can be processed further into different derivatives. The two main derivatives are chitosan and amino glucose. other applications in cosmetics include as carboxymethyl chitin (moisturizers, changes flow properties), antistatic effect due to cationic properties (hair products), as antibacterial and antifungal agents and for treatment of mucous membranes, in tumor therapies, in micro surgery, neurosurgery, for treatment of chronic wounds, ulcers and bleeding (chitin powder).

Solubility
It is soluble in concentrated formic acid and methane sulfonic acid, insoluble in water, organic solvents, weak acids and lyes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1398-61-4
Molecular Formula (C8H13NO5)nC16H28N2O11
MDL Number MFCD00466914
Synonym chitin from shrimp shells, n-2r-2,4,5-trihydroxy-6-hydroxymethyl oxan-3-yl acetamide, chitin from shrimp shells, practical grade, powder, chitin from shrimp shells, vetec tm reagent grade, chitin from shrimp shells, practical grade, coarse flakes, chitin from shrimp shells, bioreagent, suitable for analysis of chitinase, purified powder

Specifications

Color White
Quantity 100 g
Merck Index 14,2065
Solubility Information It is soluble in concentrated formic acid and methane sulfonic acid,insoluble in water,organic solvents,weak acids and lyes.
Formula Weight 209.19
Physical Form Powder
Chemical Name or Material Chitin
EINECSNumber 215-744-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.