Learn More
New England Biolabs, Inc. Chitin Resin – 20 ml
SDP
Catalog No. 50812666
Encompass
An affinity matrix for the isolation of target proteins fused to an intein-chitin binding domain fusion
Catalog No. 50-812-666 Supplier New England Biolabs, Inc. Supplier No. S6651S
An affinity matrix for the isolation of target proteins fused to an intein-chitin binding domain fusion