Learn More
Medchemexpress LLC Chitin, from crab carapace | 1398-61-4 | 25 G
SDP
Catalog No. 5000439512
Encompass_Preferred
Chitin, from crab carapace, is a long-chain polymer of N-acetylglucosamine with β-(1-4) linkages, found in the exoskeleton of crabs. It inhibits the activation of NF-κB p65, alters the translocation of NF-κB p65 to the nucleus, and interacts with the cell wall of Candida species. This product exhibits antifungal and anti-inflammatory effects and can be utilized in research concerning gastric ulcers and candidiasis.
  • Inhibits the activation of NF-κB p65.
  • Alters the translocation of NF-κB p65 to the nucleus.
  • Interacts with the cell wall of Candida species.
  • Exerts antifungal effects.
  • Exerts anti-inflammatory effects.
  • Can be used in research of gastric ulcer.
  • Can be used in research of candidiasis.
Catalog No. 50-004-39512 Supplier Medchemexpress LLC Supplier No. 25GHY126389
Chitin, from crab carapace, is a long-chain polymer of N-acetylglucosamine with β-(1-4) linkages, found in the exoskeleton of crabs. It inhibits the activation of NF-κB p65, alters the translocation of NF-κB p65 to the nucleus, and interacts with the cell wall of Candida species. This product exhibits antifungal and anti-inflammatory effects and can be utilized in research concerning gastric ulcers and candidiasis.
  • Inhibits the activation of NF-κB p65.
  • Alters the translocation of NF-κB p65 to the nucleus.
  • Interacts with the cell wall of Candida species.
  • Exerts antifungal effects.
  • Exerts anti-inflammatory effects.
  • Can be used in research of gastric ulcer.
  • Can be used in research of candidiasis.