Learn More
New England Biolabs, Inc. Chitin Magnetic Beads – 5 ml
SDP
Supplier:  New England Biolabs, Inc. E8036S
Encompass
An affinity matrix for the small-scale isolation of target proteins fused to a chitin binding domain (CBD).
Catalog No. 50-995-073