Learn More
Cusabio Technology LLC chitinase 1 chitotriosidase
Catalog No. 5000050091
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Human chitinase 1 (chitotriosidase CHIT1) in serum plasma cell culture supernates and tissue homogenates Utilizing a sandwich assay format the kit demonstrates a detection range of 31 25 pg/mL to 2000 pg/mL with a sensitivity of 7 8 pg/mL ensuring precise measurement across diverse sample types The procedure requires a modest sample volume of 50-100 L and delivers results within 1-5 hours Detection is performed at 450 nm providing reliable and reproducible data for research applications This kit is optimized for specificity and accuracy making it suitable for studying CHIT1 levels in both clinical and experimental settings
Catalog No. 50-000-50091 Supplier Cusabio Technology LLC Supplier No. CSBE17930H
This ELISA kit is designed for the quantitative detection of Human chitinase 1 (chitotriosidase CHIT1) in serum plasma cell culture supernates and tissue homogenates Utilizing a sandwich assay format the kit demonstrates a detection range of 31 25 pg/mL to 2000 pg/mL with a sensitivity of 7 8 pg/mL ensuring precise measurement across diverse sample types The procedure requires a modest sample volume of 50-100 L and delivers results within 1-5 hours Detection is performed at 450 nm providing reliable and reproducible data for research applications This kit is optimized for specificity and accuracy making it suitable for studying CHIT1 levels in both clinical and experimental settings