Learn More
ACROBiosystems ACROBiosystems Human Chitinase 3-like 1 / CHI3L1 Protein, His Tag
Supplier:  ACROBiosystems CH1H5228100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CHI3L1, His Tag (CH1-H5228) is expressed from human 293 cells (HEK293). It contains AA Tyr 22 - Thr 383 (Accession # AAH38354).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.2 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7485