Learn More
ACROBiosystems Recombinant Protein;Human Chitinase 3-like 1 / CHI3L1 Protein;HEK293;1MG;CH1,CHI3L1,YKL-40,CGP-39,GP-39,hCGP-39,ASRT7,CHI3-L1,YKL40,CGP39,GP39,hCGP39,ASRT-7,Chitinase 3-like 1
Catalog No. 501658309
Encompass_Preferred
Recombinant Protein;Human Chitinase 3-like 1 / CHI3L1 Protein;1MG;Human CHI3L1, His Tag (CH1-H5228) is expressed from human 293 cells (HEK293). It contains AA Tyr 22 - Thr 383 (Accession # AAH38354).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41 kDa. The protein migrates as 42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8309 Supplier ACROBiosystems Supplier No. CH1H52281MG
Recombinant Protein;Human Chitinase 3-like 1 / CHI3L1 Protein;1MG;Human CHI3L1, His Tag (CH1-H5228) is expressed from human 293 cells (HEK293). It contains AA Tyr 22 - Thr 383 (Accession # AAH38354).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41 kDa. The protein migrates as 42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.