Learn More
Medchemexpress LLC Chitobiose dihydrochloride | 115350-24-8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN7697B10MG
Encompass_Preferred
Chitobiose dihydrochloride is an orally active chitosan oligosaccharide with a degree of polymerization of 2. It demonstrates hepatoprotective activity and can counteract CCl4-induced elevation of plasma aspartate transaminase and alanine transaminase activities in rats, making it useful in the research of carbon tetrachloride-induced acute hepatotoxicity.
  • Orally active chitosan oligosaccharide
  • Demonstrates hepatoprotective activity
  • Can counteract CCl4-induced elevation of plasma aspartate transaminase and alanine transaminase activities
  • Used in research of carbon tetrachloride-induced acute hepatotoxicity
  • Significantly reduces CCl4-induced acute hepatotoxicity in male Wistar rats
  • Suppresses lipid peroxidation and hepatic transaminase elevation
Catalog No. 50-004-18790