Learn More
Medchemexpress LLC Chitosan | 9012-76-4 | 90.0% | 10 G
SDP
Catalog No. 5000412312
Encompass_Preferred
Chitosan (MW 30000) is a polycationic linear polysaccharide derived from chitin with an average molecular weight of 30000. It is a versatile biomaterial, recognized for its non-toxicity, low allergenicity, biocompatibility, and biodegradability. This compound also demonstrates antitumor, antibacterial, antifungal, and antioxidant activities.
  • Reduces growth potential and induces pro-apoptotic effects in certain melanoma cells.
  • Low molecular weight forms inhibit bacterial DNA transcription and mRNA synthesis.
  • High molecular weight forms alter bacterial cell wall permeability.
  • Used in water treatment, wound healing, drug delivery, obesity research, and tissue engineering.
  • Reduces aberrant crypt foci formation in colonic precancerous lesions in mice.
Catalog No. 50-004-12312 Supplier Medchemexpress LLC Supplier No. HYB2144B10G
Chitosan (MW 30000) is a polycationic linear polysaccharide derived from chitin with an average molecular weight of 30000. It is a versatile biomaterial, recognized for its non-toxicity, low allergenicity, biocompatibility, and biodegradability. This compound also demonstrates antitumor, antibacterial, antifungal, and antioxidant activities.
  • Reduces growth potential and induces pro-apoptotic effects in certain melanoma cells.
  • Low molecular weight forms inhibit bacterial DNA transcription and mRNA synthesis.
  • High molecular weight forms alter bacterial cell wall permeability.
  • Used in water treatment, wound healing, drug delivery, obesity research, and tissue engineering.
  • Reduces aberrant crypt foci formation in colonic precancerous lesions in mice.