Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitosan MP Biomedicals

Catalog No. ICN15059725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15059725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15059725 Supplier MP Biomedicals, Inc Supplier No. 0215059725
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9012-76-4
Molecular Formula C56H103N9O39
Molecular Weight (g/mol) 1526.464
InChI Key FLASNYPZGWUPSU-SICDJOISSA-N
Synonym chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f
PubChem CID 71853
IUPAC Name methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy
SMILES COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O

Specifications

Quantity 25 g
Chemical Name or Material Chitosan, From Crustacean Chitin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.