Learn More
Medchemexpress LLC HY-B2144A 1g , Chitosan (MW 150000) CAS:9012-76-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2144A1G
Encompass_Preferred
HY-B2144A 1g Chitosan (MW 150000) CAS: 9012-76-4 Chitosan (MW 150000) (Deacetylated chitin (MW 150000)) is a polycationic linear polysaccharide derived from chitin with the molecular weight of 150000. Chitosan is an versatile biomaterial because of its non-toxicity, low allergenicity, biocompatibility and biodegradability. Chitosan also has antitumor, antibacterial, antifungal, and antioxidant activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1934