Learn More
Medchemexpress LLC Chitosan (MW 150000) | 9012-76-4 | 10 G
SDP
Catalog No. 5000410920
Encompass_Preferred
Chitosan (MW 150000) is a polycationic linear polysaccharide derived from chitin. This versatile biomaterial is recognized for its non-toxicity, low allergenicity, biocompatibility, and biodegradability. It also exhibits antitumor, antibacterial, antifungal, and antioxidant properties.
  • Derived from chitin
  • Polycationic linear polysaccharide
  • Non-toxic and low allergenicity
  • Biocompatible and biodegradable
  • Exhibits antitumor, antibacterial, antifungal, and antioxidant activities
  • Can be utilized in water treatment and wound-healing materials
  • Suitable as a pharmaceutical excipient or drug carrier
  • Used in obesity research and as a scaffold for tissue engineering
Catalog No. 50-004-10920 Supplier Medchemexpress LLC Supplier No. HYB2144A10G
Chitosan (MW 150000) is a polycationic linear polysaccharide derived from chitin. This versatile biomaterial is recognized for its non-toxicity, low allergenicity, biocompatibility, and biodegradability. It also exhibits antitumor, antibacterial, antifungal, and antioxidant properties.
  • Derived from chitin
  • Polycationic linear polysaccharide
  • Non-toxic and low allergenicity
  • Biocompatible and biodegradable
  • Exhibits antitumor, antibacterial, antifungal, and antioxidant activities
  • Can be utilized in water treatment and wound-healing materials
  • Suitable as a pharmaceutical excipient or drug carrier
  • Used in obesity research and as a scaffold for tissue engineering